Assay Details
| Gene Symbol: ATP8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105529.1 | NP_001098999.1 | 5 - 6 | 645 | 145 | |
| XM_005248043.2 | XP_005248100.1 | 5 - 6 | 640 | 145 | ||
| XM_011513615.1 | XP_011511917.1 | 5 - 6 | 640 | 145 | ||
| XM_011513618.1 | XP_011511920.1 | 5 - 6 | 640 | 145 | ||
| GenBank mRNA | AB209687.1 | - | 5 - 6 | 498 | 145 | |
| BC109317.1 | - | 5 - 6 | 407 | 40036536 | 145 | |
| BC109318.1 | - | 5 - 6 | 413 | 40036538 | 145 | |
More Information
Set Membership: |
|

ATP8A1
Genomic Map