Assay Details
| Gene Symbol: CC2D2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011539789.2 | XP_011538091.1 | 2 - 3 | 263 | 127 | |
| XM_011539790.2 | XP_011538092.1 | 2 - 3 | 263 | 127 | ||
| XM_011539791.2 | XP_011538093.1 | 2 - 3 | 263 | 127 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000484685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000484685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012484.hg.1 | ENST00000484685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000469549.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000472454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000484685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000484685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012484.hg.1 | ENST00000484685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012486.hg.1 | ENST00000475252.2 | Ensembl |

CC2D2B
Genomic Map