Assay Details
Gene Symbol: DHX58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024119.2 | NP_077024.2 | 3 - 4 | 396 | 66 | |
XM_017025059.1 | XP_016880548.1 | 2 - 3 | 329 | 66 | ||
XM_017025060.1 | XP_016880549.1 | 3 - 4 | 392 | 66 | ||
GenBank mRNA | AK021416.1 | - | 3 - 4 | 396 | 66 | |
AK097669.1 | - | 2 - 3 | 339 | 66 | ||
AK225549.1 | - | 3 - 4 | 396 | 66 | ||
BC014949.1 | - | 3 - 4 | 333 | 4865798 | 66 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001523.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to virus
negative regulation of type I interferon production
positive regulation of type I interferon production
negative regulation of MDA-5 signaling pathway
negative regulation of RIG-I signaling pathway
innate immune response
regulation of innate immune response
negative regulation of innate immune response
defense response to virus
positive regulation of MDA-5 signaling pathway
positive regulation of RIG-I signaling pathway
DNA binding
double-stranded RNA binding
single-stranded RNA binding
helicase activity
protein binding
ATP binding
zinc ion binding
response to virus
negative regulation of type I interferon production
positive regulation of type I interferon production
negative regulation of MDA-5 signaling pathway
negative regulation of RIG-I signaling pathway
innate immune response
regulation of innate immune response
negative regulation of innate immune response
defense response to virus
positive regulation of MDA-5 signaling pathway
positive regulation of RIG-I signaling pathway
DNA binding
double-stranded RNA binding
single-stranded RNA binding
helicase activity
protein binding
ATP binding
zinc ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17020299.hg.1 | ENST00000251642.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17020299.hg.1 | ENST00000423748.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17020299.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17020299.hg.1 | TC17001523.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000430773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000430773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000430773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000430773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000430773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000589979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000589979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000589979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000589979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000589979.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000590637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000590637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000590637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000590637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000590637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000590637.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000592024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000592024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000592024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | ENST00000592024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | ENST00000430773.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | ENST00000430773.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700184966.hg.1 | TC1700010707.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000251642.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000413196.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000430773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000430773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000430773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000430773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000430773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000586522.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000589979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000589979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000589979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000589979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000589979.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000590637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000590637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000590637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000590637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000590637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000590637.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000591220.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000592024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000592024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000592024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | ENST00000592024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010707.hg.1 | NM_024119.2 | Refseq |
