Assay Details
| Gene Symbol: OLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172632.1 | NP_001166103.1 | 3 - 4 | 531 | 133 | |
| GenBank mRNA | AK295409.1 | - | 3 - 4 | 472 | 133 | |
| AK316353.1 | - | 2 - 3 | 261 | 133 | ||
More Information
Set Membership: |
|

OLR1
Genomic Map