Assay Details
| Gene Symbol: PTEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000314.6 | NP_000305.3 | 9 | 3334 | 137 | |
| NM_001304717.2 | NP_001291646.2 | 9 | 3334 | 137 | ||
| NM_001304718.1 | NP_001291647.1 | 9 | 3448 | 137 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

PTEN
Genomic Map