Assay Details
| Gene Symbol: MZT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005263741.4 | XP_005263798.1 | 3 | 916 | 84 | |
| GenBank mRNA | AK090531.1 | - | 1 | 1455 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000281871.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000281871.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000281871.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000281871.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000281871.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000281871.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000281871.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000281871.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000281871.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000281871.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000409255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000409255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000409255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000409255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000409255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000409255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000409255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000409255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000409255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000425361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000425361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000425361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000425361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000425361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000425361.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000455239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000455239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000455239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000455239.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000457492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000457492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000457492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000457492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000457492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000457492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000480182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000480182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000480182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000480182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000480182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000485869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000485869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000485869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000485869.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000491178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | ENST00000491178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | NM_025029.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | NM_025029.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | NM_025029.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | NM_025029.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | NM_025029.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | NM_025029.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | NM_025029.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | NM_025029.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | NM_025029.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009281.hg.1 | NM_025029.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009282.hg.1 | ENST00000578074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000309451.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000309451.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000309451.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000309451.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000309451.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000309451.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000410036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000410036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000410036.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000488586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000488586.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000491265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000491265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000491265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000491265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000491265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000491265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | ENST00000579560.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | NM_001085365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | NM_001085365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | NM_001085365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | NM_001085365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | NM_001085365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | NM_001085365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | NM_001085365.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016732.hg.1 | NR_039945.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009282.hg.1 | ENST00000578074.1 | Ensembl |

MZT2A
Genomic Map