| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
ENST00000602032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
ENST00000602032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900192175.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192175.hg.1 |
ENST00000602032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192175.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900192175.hg.1 |
TC1900011056.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900192175.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192175.hg.1 |
ENST00000602032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192175.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900192173.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192173.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900192173.hg.1 |
TC1900011056.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900192173.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192173.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900192172.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192172.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900192172.hg.1 |
TC1900011056.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900192172.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192172.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900192178.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192178.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900192178.hg.1 |
TC1900011056.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900192178.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192178.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900192177.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192177.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900192177.hg.1 |
TC1900011056.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900192177.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900192177.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
ENST00000293255.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
ENST00000602032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
ENST00000602032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011056.hg.1 |
NM_019855.4 |
Refseq |