| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021177.hg.1 |
ENST00000321702.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021177.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021177.hg.1 |
TC19001556.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021175.hg.1 |
ENST00000321702.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021175.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021175.hg.1 |
TC19001556.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000601379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000601379.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000601616.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
ENST00000601616.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900188869.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900188869.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900188869.hg.1 |
TC1900010747.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900188869.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900188869.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900188871.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900188871.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900188871.hg.1 |
TC1900010747.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900188871.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900188871.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900188872.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900188872.hg.1 |
ENST00000601379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900188872.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900188872.hg.1 |
TC1900010747.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900188872.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900188872.hg.1 |
ENST00000601379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900188872.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900188868.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900188868.hg.1 |
ENST00000601379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900188868.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900188868.hg.1 |
TC1900010747.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900188868.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900188868.hg.1 |
ENST00000601379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900188868.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000321702.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000601379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000601379.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000601616.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
ENST00000601616.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010747.hg.1 |
NM_198540.2 |
Refseq |