Assay Details
Gene Symbol: DMBT1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003570.2 | 14 - 15 | 1552 | 62 | ||
GenBank mRNA | AK128228.1 | - | 14 - 15 | 1552 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromosome 10 open reading frame 120 | C10orf120 | bA318C4.1 | -77.9kb | Anti-sense | Chr. 10 - 122697709 - 122699822 on NCBI Build GRCh38 |
Downstream | CUB and zona pellucida like domains 1 | CUZD1 |
ERG-1, ERG1, ITMAP1, UO-44 |
52.8kb | Anti-sense | Chr. 10 - 122832155 - 122846175 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000892.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK128228.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10010922.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10010922.hg.1 | TC10000892.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162820.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162820.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162820.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162820.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162820.hg.1 | TC1000009159.hg.1 | Clariom D TC |
