Assay Details
Gene Symbol: DMBT1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003570.2 | 24 - 25 | 2925 | 81 | ||
GenBank mRNA | AK128228.1 | - | 24 - 25 | 2925 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromosome 10 open reading frame 120 | C10orf120 | bA318C4.1 | -92.5kb | Anti-sense | Chr. 10 - 122697709 - 122699822 on NCBI Build GRCh38 |
Downstream | deleted in malignant brain tumors 1 protein | LOC729800 | 6.2kb | Sense | Chr. 10 - 122800976 - 122826868 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000892.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10010929.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10010929.hg.1 | TC10000892.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10010931.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10010931.hg.1 | TC10000892.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10010930.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10010930.hg.1 | TC10000892.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10010932.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10010932.hg.1 | TC10000892.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162830.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162830.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162830.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162830.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162830.hg.1 | TC1000009159.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000162829.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162829.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162829.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162829.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162829.hg.1 | TC1000009159.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000162831.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162831.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162831.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162831.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162831.hg.1 | TC1000009159.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000605982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | ENST00000607497.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009159.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162832.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162832.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162832.hg.1 | ENST00000439464.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000162832.hg.1 | NR_003570.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000162832.hg.1 | TC1000009159.hg.1 | Clariom D TC |
