Assay Details
Gene Symbol: FAM212B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198926.2 | NP_945120.1 | 1 - 2 | 95 | 81 | |
GenBank mRNA | AK293989.1 | - | 1 - 2 | 122 | 81 | |
AL137199.1 | - | 1 - 2 | 95 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002990.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK293989.1,AL137199.1,NM_198926.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01046416.hg.1 | TC01002990.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01046416.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01046416.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01046416.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01046417.hg.1 | TC01002990.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01046417.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01046417.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01046417.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01046417.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01046423.hg.1 | TC01002990.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01046423.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01046423.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213969.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213969.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213969.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213969.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213969.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213969.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213969.hg.1 | TC0100015284.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100213971.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213971.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213971.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213971.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213971.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213971.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213971.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213971.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213971.hg.1 | TC0100015284.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100213972.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213972.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213972.hg.1 | ENST00000527621.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213972.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213972.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213972.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213972.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213972.hg.1 | ENST00000527621.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213972.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213972.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213972.hg.1 | TC0100015284.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000527621.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | ENST00000534365.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015284.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213982.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213982.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213982.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213982.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213982.hg.1 | TC0100015284.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100213970.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213970.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213970.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213970.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213970.hg.1 | ENST00000357260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213970.hg.1 | ENST00000444059.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100213970.hg.1 | NM_019099.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213970.hg.1 | NM_198926.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100213970.hg.1 | TC0100015284.hg.1 | Clariom D TC |
