Assay Details
| Gene Symbol: CYP4V2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK122600.1 | - | 6 - 7 | 841 | 149 | |
| AK126473.1 | - | 7 - 8 | 775 | 149 | ||
| AY422002.1 | - | 6 - 7 | 1106 | 149 | ||
| BC060857.1 | - | 6 - 7 | 972 | 30333559 | 149 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400189397.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189397.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189397.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400189397.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189397.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189397.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400189397.hg.1 | TC0400012874.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000502665.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | ENST00000513354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012874.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400189396.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189396.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189396.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400189396.hg.1 | ENST00000378802.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189396.hg.1 | ENST00000507209.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400189396.hg.1 | NM_207352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400189396.hg.1 | TC0400012874.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 104577 | - | - |

CYP4V2
Genomic Map