Assay Details
Gene Symbol: LINC00299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK127578.1 | - | 4 - 5 | 570 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000454043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000454043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000454043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000454043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000454043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000454043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
