Assay Details
Gene Symbol: LINC00299 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK127578.1 | - | 5 - 6 | 964 | 114 | |
BC043563.1 | - | 3 - 4 | 310 | 114 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 298 | LINC00298 | -318.0kb | Sense | Chr. 2 - 7922425 - 7976814 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101929567 | LOC101929567 | 262.9kb | Sense | Chr. 2 - 8559833 - 8583792 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000454043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000454043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | ENST00000454043.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000430192.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000442956.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000454043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000454043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | ENST00000454043.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016621.hg.1 | NR_034135.1 | Refseq |
