Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
7937699 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
7937698 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006523.hg.1 |
ENST00000624434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006523.hg.1 |
ENST00000624434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006523.hg.1 |
ENST00000624434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006523.hg.1 |
ENST00000624434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
ENST00000382171.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100009745.hg.1 |
NM_001005922.1 |
Refseq |