Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8021775 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8021776 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8021774 |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007722.hg.1 |
ENST00000622985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007722.hg.1 |
ENST00000622985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007722.hg.1 |
NM_001272093.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007722.hg.1 |
NM_001272093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152804.hg.1 |
ENST00000622985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152804.hg.1 |
NM_001272093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152804.hg.1 |
TC1800007722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152804.hg.1 |
ENST00000622985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152804.hg.1 |
NM_001272093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007722.hg.1 |
ENST00000622985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007722.hg.1 |
ENST00000622985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007722.hg.1 |
NM_001272093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007722.hg.1 |
NM_001272093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152803.hg.1 |
ENST00000622985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152803.hg.1 |
NM_001272093.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800152803.hg.1 |
TC1800007722.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800152803.hg.1 |
ENST00000622985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800152803.hg.1 |
NM_001272093.2 |
Refseq |