Assay Details
Gene Symbol: FLJ46365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK128232.1 | - | 1 - 2 | 297 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101929268 | LOC101929268 | Sense | Chr. 8 - 48551567 - 48698510 on NCBI Build GRCh38 | ||
Downstream | uncharacterized LOC101929217 | LOC101929217 | 28.9kb | Sense | Chr. 8 - 48620403 - 48623895 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388476 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104959 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388497 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388507 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388478 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104963 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388525 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104957 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104960 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104955 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104954 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388475 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104937 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388508 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104966 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388440 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104944 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104972 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104906 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16092853 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388506 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388451 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388489 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388487 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104913 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104895 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104925 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104923 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104973 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104951 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388479 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388449 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104939 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388457 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388491 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104907 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388492 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104909 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388529 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104896 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104962 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388505 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388474 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16092852 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388448 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388500 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388445 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104926 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388471 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16092833 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104958 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16092832 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388443 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104953 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388530 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388473 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104905 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104891 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104903 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388495 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16092840 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104941 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16092842 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104924 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388504 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104911 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388480 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388517 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388477 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104940 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388513 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P06104970 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P26388486 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3329939 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 18363 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | XR_040285_at | - | - |
Affymetrix Human Gene 1.0 ST Array | XR_040286_at | - | - |
Affymetrix Human Gene 1.0 ST Array | XR_040287_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8146389 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8146390 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8146388 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0800007568.hg.1 | ENST00000430626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007568.hg.1 | ENST00000430626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007568.hg.1 | ENST00000430626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007568.hg.1 | ENST00000430626.1 | Ensembl |
