Assay Details
| Gene Symbol: RPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000988.3 | NP_000979.1 | 3 - 4 | 295 | 155 | |
| GenBank mRNA | AK124148.1 | - | 1 - 2 | 1682 | 155 | |
| AK311792.1 | - | 3 - 4 | 285 | 155 | ||
| BC001700.2 | - | 3 - 4 | 318 | 3050183 | 155 | |
| BC002588.2 | - | 3 - 4 | 275 | 3161438 | 155 | |
| BC007273.1 | - | 3 - 4 | 318 | 3161124 | 155 | |
| BC010026.2 | - | 3 - 4 | 322 | 3354227 | 155 | |
| BC021886.2 | - | 282 | 155 | |||
| BC098560.1 | - | 290 | 30415908 | 155 | ||
| L05094.1 | - | 2 - 3 | 275 | 155 | ||
| L19527.1 | - | 2 - 3 | 267 | 155 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RPL27
Genomic Map