Assay Details
| Gene Symbol: PGM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021965.3 | NP_068800.2 | 8 - 9 | 1520 | 68 | |
| XM_017014808.1 | XP_016870297.1 | 8 - 9 | 1412 | 68 | ||
| GenBank mRNA | AK297108.1 | - | 8 - 9 | 1342 | 68 | |
| AL710966.1 | - | 3 - 4 | 324 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000312.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09003208.hg.1 | TC09000312.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09003208.hg.1 | ENST00000377314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09003208.hg.1 | ENST00000396396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09003208.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09003208.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000431583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000431583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000431583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000431583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000496758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000496758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000496758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000496758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | ENST00000496758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000431583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000431583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000431583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000431583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000496758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000496758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000496758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000496758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | ENST00000496758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012139.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184136.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184136.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184136.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184136.hg.1 | ENST00000396396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184136.hg.1 | ENST00000472639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900184136.hg.1 | NM_021965.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900184136.hg.1 | TC0900012139.hg.1 | Clariom D TC |

PGM5
Genomic Map