Assay Details
Gene Symbol: KRTAP9-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318227.1 | NP_001305156.1 | 2 | 838 | 136 | |
NM_030975.2 | NP_112237.2 | 1 | 823 | 136 | ||
GenBank mRNA | AJ406949.1 | - | 1 | 649 | 136 | |
AJ406951.1 | - | 3 | 778 | 136 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000508.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321742 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893609 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893646 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893611 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893615 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321747 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893577 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893656 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321763 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16981750 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893575 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321761 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16981755 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321743 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321745 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893576 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893645 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893668 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321753 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893574 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893640 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321757 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321738 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893651 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893659 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321741 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16981753 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16981679 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321755 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893650 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893635 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321760 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321764 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16981676 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321739 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893643 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893652 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893669 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16981746 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321716 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321756 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321759 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893638 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 46950 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 58529 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 173221 | AK091949.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 173221 | AK093376.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 173221 | AX748086.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 173221 | BC016997.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 173221 | BC032758.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 173221 | BC039051.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 173221 | BC062318.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 173221 | BC107693.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 173221 | NM_005559.2 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 22909 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 21036 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 30383 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 325455 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 162456 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 135645 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 934106 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 7793 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 26191 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | AF004230.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | AK000727.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | AK027009.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | AK098118.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | AK126272.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | AK128595.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | AK130127.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | AK130149.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | BC004147.2 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | BC015473.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | BC017840.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | CR607267.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | CR615077.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | J03801.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | M19045.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 117425 | NM_000239.1 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 367587 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 155629 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 300826 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 357542 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 187115 | AK000004.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 187115 | AK225843.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 187115 | AK292676.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 187115 | AY211386.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 187115 | BC032429.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 187115 | BC111054.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 187115 | BX647829.1 | genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 187115 | NM_001083536.1 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 187115 | NM_033086.2 | refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 336913 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 20940 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 115856 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 306882 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 125546 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 27993 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 139767 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 157695 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 225813 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 218009 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 292554 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 72219 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P311585 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 5911 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P117775 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P103891 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P103891 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P117775 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 62363 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P12648287 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P311585 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P311585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893574 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321716 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321739 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321738 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16981676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893609 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16981746 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321738 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893609 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16981676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893577 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893574 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321716 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321739 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16981746 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P117775 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P103891 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20646984 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03244863 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P103891 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P117775 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P124311 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P103891 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P117775 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P311585 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 26669 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 3527 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 128920 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 10539 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 88285 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12646628 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20646984 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P124311 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03244862 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03244863 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12648287 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P117775 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P20863546 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P103891 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321762 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321766 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321760 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321716 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321752 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321764 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321757 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321722 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321728 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321755 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321750 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321748 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321733 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321725 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321740 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321741 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321758 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321735 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321744 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321736 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321731 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321738 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321753 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321765 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321746 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321745 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P20646984 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P117775 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P103891 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03244863 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P124311 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8007124 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8007136 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8007126 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8007138 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8007135 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8007132 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8007123 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8007134 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8007130 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 220972_PM_s_at | - | - |
Affymetrix Human Genome U219 Array | 11735982_at | - | - |
Affymetrix Human Genome U219 Array | 11735982_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR17006106.hg.1 | ENST00000394008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17006106.hg.1 | NM_030975.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17006106.hg.1 | TC17000508.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700012256.hg.1 | ENST00000394008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012256.hg.1 | ENST00000394008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012256.hg.1 | NM_030975.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012256.hg.1 | NM_030975.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700202344.hg.1 | ENST00000394008.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700202344.hg.1 | NM_030975.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700202344.hg.1 | ENST00000394008.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700202344.hg.1 | NM_030975.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700202344.hg.1 | TC1700012256.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700202343.hg.1 | ENST00000394008.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700202343.hg.1 | NM_030975.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700202343.hg.1 | ENST00000394008.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700202343.hg.1 | NM_030975.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700202343.hg.1 | TC1700012256.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700012256.hg.1 | ENST00000394008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012256.hg.1 | ENST00000394008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012256.hg.1 | NM_030975.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012256.hg.1 | NM_030975.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 220972_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 220972_s_at | - | - |
Human Genome U133A 2.0 Array | 220972_s_at | - | - |
Human Genome U133A 2.0 Array | 220972_PM_s_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw526331 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220972_s_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220972_s_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220972_s_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220972_s_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220972_s_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220972_s_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220972_s_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220972_s_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220972_s_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220972_s_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220972_s_at6 | - | - |
Human Genome U133 Set | 220972_PM_s_at | - | - |
Human Genome U133 Set | 220972_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009509 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009509 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 112285 | - | - |
