Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010362.hg.1 |
ENST00000445535.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010362.hg.1 |
NR_024151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010362.hg.1 |
NR_024151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010362.hg.1 |
NR_024151.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021994.hg.1 |
TC01001401.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021994.hg.1 |
ENST00000309758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021994.hg.1 |
ENST00000545155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021994.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021993.hg.1 |
TC01001401.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021993.hg.1 |
ENST00000309758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021993.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021989.hg.1 |
TC01001401.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021989.hg.1 |
ENST00000309758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021989.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021992.hg.1 |
TC01001401.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021992.hg.1 |
ENST00000309758.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021992.hg.1 |
ENST00000545155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01021992.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022055.hg.1 |
TC01001407.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01022055.hg.1 |
NR_024151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176295.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176295.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176295.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176295.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176295.hg.1 |
TC0100010360.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100176304.hg.1 |
NR_024151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176304.hg.1 |
NR_024151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176304.hg.1 |
TC0100010362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100176305.hg.1 |
ENST00000445535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176305.hg.1 |
NR_024151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176305.hg.1 |
ENST00000445535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176305.hg.1 |
NR_024151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176305.hg.1 |
TC0100010362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010362.hg.1 |
ENST00000445535.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010362.hg.1 |
NR_024151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010362.hg.1 |
NR_024151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010362.hg.1 |
NR_024151.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176296.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176296.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176296.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100176296.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100176296.hg.1 |
TC0100010360.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
ENST00000309758.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010360.hg.1 |
NM_002155.4 |
Refseq |