Assay Details
Gene Symbol: SMIM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320261.1 | NP_001307190.1 | 1 - 2 | 164 | 137 | |
GenBank mRNA | BI759434.1 | - | 1 - 2 | 104 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001320261.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
