Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002664.hg.1 |
TC05003439.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002664.hg.1 |
ENST00000282588.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002664.hg.1 |
NM_181501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500149769.hg.1 |
ENST00000282588.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500149769.hg.1 |
ENST00000504669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500149769.hg.1 |
ENST00000506275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500149769.hg.1 |
NM_181501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500149769.hg.1 |
ENST00000282588.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500149769.hg.1 |
ENST00000504669.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500149769.hg.1 |
ENST00000506275.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500149769.hg.1 |
NM_181501.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500149769.hg.1 |
TC0500007365.hg.1 |
Clariom D TC |