Assay Details
Gene Symbol: C9orf153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276366.1 | NP_001263295.1 | 2 - 3 | 197 | 124 | |
NM_001276367.1 | NP_001263296.1 | 3 - 4 | 276 | 124 | ||
NM_001276368.1 | NP_001263297.1 | 3 - 4 | 215 | 124 | ||
GenBank mRNA | BC108665.1 | - | 3 - 4 | 254 | 5164660 | 124 |
BG772848.1 | - | 2 - 3 | 202 | 124 | ||
BI827110.1 | - | 3 - 4 | 216 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000376001.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000376001.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000376001.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000376001.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000376001.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000469914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000469914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000469914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000469914.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000617985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000617985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000617985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | ENST00000617985.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | ENST00000376001.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | ENST00000617985.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | ENST00000376001.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | ENST00000617985.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172491.hg.1 | TC0900010618.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000339137.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000376001.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000376001.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000376001.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000376001.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000376001.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000469914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000469914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000469914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000469914.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000613665.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000617985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000617985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000617985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | ENST00000617985.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276366.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276367.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010618.hg.1 | NM_001276368.1 | Refseq |
