Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012938.hg.1 |
ENST00000434887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012938.hg.1 |
ENST00000434887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012938.hg.1 |
ENST00000434887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012938.hg.1 |
ENST00000434887.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012938.hg.1 |
NR_110829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012938.hg.1 |
NR_110829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012938.hg.1 |
NR_110829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012938.hg.1 |
NR_110829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012938.hg.1 |
ENST00000434887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012938.hg.1 |
ENST00000434887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012938.hg.1 |
ENST00000434887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012938.hg.1 |
ENST00000434887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012938.hg.1 |
NR_110829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012938.hg.1 |
NR_110829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012938.hg.1 |
NR_110829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012938.hg.1 |
NR_110829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193339.hg.1 |
ENST00000434887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193339.hg.1 |
NR_110829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193339.hg.1 |
ENST00000434887.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700193339.hg.1 |
NR_110829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700193339.hg.1 |
TC0700012938.hg.1 |
Clariom D TC |