Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900006619.hg.1 |
ENST00000391389.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007036.hg.1 |
ENST00000483614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007036.hg.1 |
ENST00000483614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007036.hg.1 |
ENST00000483614.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012518.hg.1 |
ENST00000456046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012518.hg.1 |
ENST00000456046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012518.hg.1 |
ENST00000456046.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200012518.hg.1 |
ENST00000456046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007036.hg.1 |
ENST00000483614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007036.hg.1 |
ENST00000483614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007036.hg.1 |
ENST00000483614.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012518.hg.1 |
ENST00000456046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012518.hg.1 |
ENST00000456046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012518.hg.1 |
ENST00000456046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200012518.hg.1 |
ENST00000456046.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900006619.hg.1 |
ENST00000391389.3 |
Ensembl |