Assay Details
| Gene Symbol: ZNF253 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021047.2 | NP_066385.2 | 3 - 4 | 318 | 141 | |
| GenBank mRNA | AF038951.1 | - | 2 - 3 | 226 | 141 | |
| AK074757.1 | - | 3 - 4 | 310 | 141 | ||
| BC014148.2 | - | 3 - 4 | 318 | 141 | ||
| BC125063.1 | - | 3 - 4 | 297 | 40117279 | 141 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Ambion siRNA available  Amplicon length greater than or equal to 101  Mammalian Gene Collection  Homo sapiens  Probe spans exons | 
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 ZNF253
ZNF253 Genomic Map
Genomic Map Back To Top
 Back To Top