Assay Details
| Gene Symbol: MRPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300848.1 | NP_001287777.1 | 1 - 2 | 260 | 146 | |
| NM_015950.4 | NP_057034.2 | 1 - 2 | 260 | 146 | ||
| GenBank mRNA | AF086160.1 | - | 1 - 2 | 120 | 146 | |
| AF132956.1 | - | 1 - 2 | 134 | 146 | ||
| BM563754.1 | - | 226 | 146 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF132956.1, NM_001300848.1, NM_015950.4 transcripts.
Set Membership: |
|

MRPL2
Genomic Map