Assay Details
| Gene Symbol: MRPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300848.1 | NP_001287777.1 | 4 - 5 | 685 | 118 | |
| NM_015950.4 | NP_057034.2 | 4 - 5 | 685 | 118 | ||
| XM_005249161.4 | XP_005249218.1 | 4 - 5 | 506 | 118 | ||
| GenBank mRNA | AF132956.1 | - | 4 - 5 | 559 | 118 | |
| AK314948.1 | - | 4 - 5 | 543 | 118 | ||
| BC013685.1 | - | 4 - 5 | 530 | 3862065 | 118 | |
| BC020212.1 | - | 4 - 5 | 530 | 4641018 | 118 | |
| BM563754.1 | - | 651 | 118 | |||
| BM970997.1 | - | 2 - 3 | 118 | |||
More Information
Set Membership: |
|

MRPL2
Genomic Map