| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100007280.hg.1 |
ENST00000357130.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000079.hg.1 |
TC04000006.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000079.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000079.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000079.hg.1 |
ENST00000505939.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000079.hg.1 |
ENST00000512994.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000079.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013225.hg.1 |
ENST00000418060.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000078.hg.1 |
TC04000006.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000078.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000078.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000078.hg.1 |
ENST00000505939.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000078.hg.1 |
ENST00000512994.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000078.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000080.hg.1 |
TC04000006.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000080.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000080.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000080.hg.1 |
ENST00000505939.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000080.hg.1 |
ENST00000512994.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04000080.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008845.hg.1 |
ENST00000440423.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012882.hg.1 |
ENST00000419098.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012882.hg.1 |
ENST00000419098.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012882.hg.1 |
ENST00000419098.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012882.hg.1 |
ENST00000419098.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012882.hg.1 |
ENST00000619749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012882.hg.1 |
ENST00000619749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012882.hg.1 |
ENST00000619749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012882.hg.1 |
ENST00000619749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012882.hg.1 |
NM_001137608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012882.hg.1 |
NM_001137608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012882.hg.1 |
NM_001137608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012882.hg.1 |
NM_001137608.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013854.hg.1 |
ENST00000420753.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006789.hg.1 |
ENST00000402970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006789.hg.1 |
ENST00000402970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008845.hg.1 |
ENST00000440423.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100007280.hg.1 |
ENST00000357130.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012882.hg.1 |
ENST00000419098.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012882.hg.1 |
ENST00000419098.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012882.hg.1 |
ENST00000419098.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012882.hg.1 |
ENST00000419098.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012882.hg.1 |
ENST00000619749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012882.hg.1 |
ENST00000619749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012882.hg.1 |
ENST00000619749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012882.hg.1 |
ENST00000619749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012882.hg.1 |
NM_001137608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012882.hg.1 |
NM_001137608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012882.hg.1 |
NM_001137608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012882.hg.1 |
NM_001137608.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400187587.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187587.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187587.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400187587.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187587.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187587.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400187587.hg.1 |
TC0400012754.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400187584.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187584.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187584.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187584.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400187584.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187584.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187584.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187584.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400187584.hg.1 |
TC0400012754.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200013225.hg.1 |
ENST00000418060.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013854.hg.1 |
ENST00000420753.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187585.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187585.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187585.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187585.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400187585.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187585.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187585.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400187585.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400187585.hg.1 |
TC0400012754.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000240499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000366506.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
ENST00000503699.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012754.hg.1 |
NM_003441.3 |
Refseq |