Assay Details
Gene Symbol: PYY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003064.2 | 2 | 695 | 128 | ||
GenBank mRNA | AF222904.1 | - | 2 | 444 | 128 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105371930 | LOC105371930 | Anti-sense | Chr. 17 - 28222783 - 28228461 on NCBI Build GRCh38 | ||
Downstream | uncharacterized LOC105371712 | LOC105371712 | 18.4kb | Anti-sense | Chr. 17 - 28246454 - 28250825 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10271263 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30835247 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10271260 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10271264 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10271270 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30835243 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10271271 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16965183 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3257866 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P233078 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P233078 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 38463 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4434 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 39738 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P233078 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3257866 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P233078 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3257866 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271251 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835234 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965196 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271258 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835237 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835239 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271272 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835224 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835243 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965183 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965186 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835222 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835256 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271257 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835225 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271252 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835247 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271263 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271264 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271256 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271246 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835257 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271254 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835236 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271259 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835227 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271260 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271271 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271265 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965193 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271271 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835247 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835243 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965186 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835234 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271265 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271254 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271258 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965196 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835239 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835237 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835257 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835256 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271259 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271251 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271264 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271272 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835227 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835236 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835224 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271260 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271263 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271246 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271256 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835225 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271252 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30835222 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10271257 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965193 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16965183 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P233078 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P233078 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271270 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271272 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271275 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271271 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271277 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16965199 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271258 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271256 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271250 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16965167 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16965195 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271268 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271264 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271267 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271269 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271252 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271254 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271265 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16965186 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271248 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271274 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10271260 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_003064_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8005834 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8005835 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8005833 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 220209_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11744736_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11744736_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000315776.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000315776.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000315776.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000315776.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007355.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152258.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152258.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152258.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152258.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152258.hg.1 | TC1700007355.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700152259.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152259.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152259.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152259.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152259.hg.1 | TC1700007355.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700152257.hg.1 | ENST00000315776.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152257.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152257.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152257.hg.1 | ENST00000315776.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152257.hg.1 | ENST00000441253.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152257.hg.1 | NR_003064.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152257.hg.1 | TC1700007355.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 220209_at | - | - |
Human Genome U133 Plus 2.0 Array | 220209_PM_at | - | - |
Human Genome U133A 2.0 Array | 220209_at | - | - |
Human Genome U133A 2.0 Array | 220209_PM_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw523903 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 220209_at1 | - | - |
Human Genome U95 Set | 83732_at | - | - |
Human Genome U95 Set | 83732_PM_at | - | - |
Human Genome U133 Set | 220209_at | - | - |
Human Genome U133 Set | 220209_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009296 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009295 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009296 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009295 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 212372 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1699565 | AF222904.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1699565 | AF222904.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1699565 | AF222904.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1699565 | AF222904.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1699565 | AF222904.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1699565 | AF222904.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2070395 | NR_003064.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1699565 | AF222904.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2070395 | NR_003064.2 | human_transcriptome |
