Assay Details
Gene Symbol: LRRC37A7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC036040.1 | - | 1 | 1374 | 175 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | beta-1,4-galactosyltransferase 6 | B4GALT6 |
B4Gal-T6, beta4Gal-T6 |
Anti-sense | Chr. 18 - 31622246 - 31724670 on NCBI Build GRCh38 | |
Downstream | solute carrier family 25 member 52 | SLC25A52 | MCART2 | 32.7kb | Anti-sense | Chr. 18 - 31759696 - 31760880 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100007683.hg.1 | ENST00000604085.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007021.hg.1 | ENST00000583900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007021.hg.1 | ENST00000583900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007021.hg.1 | ENST00000583900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007021.hg.1 | ENST00000583900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007021.hg.1 | ENST00000583900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007021.hg.1 | ENST00000585232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007021.hg.1 | ENST00000585232.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013614.hg.1 | ENST00000454917.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013614.hg.1 | ENST00000454917.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013614.hg.1 | ENST00000454917.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013614.hg.1 | ENST00000454917.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007021.hg.1 | ENST00000583900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007021.hg.1 | ENST00000583900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007021.hg.1 | ENST00000583900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007021.hg.1 | ENST00000583900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007021.hg.1 | ENST00000583900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007021.hg.1 | ENST00000585232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007021.hg.1 | ENST00000585232.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007683.hg.1 | ENST00000604085.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000425901.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | ENST00000579258.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007730.hg.1 | NR_033753.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013614.hg.1 | ENST00000454917.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013614.hg.1 | ENST00000454917.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013614.hg.1 | ENST00000454917.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013614.hg.1 | ENST00000454917.1 | Ensembl |
