Assay Details
Gene Symbol: CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204477.1 | NP_001191406.1 | 4 | 2436 | 153 | |
GenBank mRNA | AK093993.1 | - | 2 | 2180 | 153 | |
AK172733.1 | - | 1 | 1573 | 153 | ||
BM971397.1 | - | 1 | 153 |
Gene Symbol: TVP23C-CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204478.1 | NP_001191407.1 | 7 | 2847 | 153 | |
NR_037924.1 | 6 | 2702 | 153 | |||
GenBank mRNA | AB074181.1 | - | 7 | 2777 | 153 | |
CB850543.1 | - | 1 | 153 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10249499 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30800906 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30800908 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30800905 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16946445 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10249498 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30800907 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16946447 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30800900 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10249493 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10249494 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30800897 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P262321 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P262321 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P427083 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P262321 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P40764223 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03220508 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P40764223 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P20597515 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 16000 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 160682 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 80231 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20597542 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20597537 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12597076 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03220495 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40764223 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03220504 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03220482 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20597515 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40764286 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20597556 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03220485 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20597579 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40764273 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03220500 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P20597527 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03220508 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12596117 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03220499 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8012925 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8012921 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8012920 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8012927 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8012926 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8012920 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8012922 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8012923 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8012918 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8012918 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 244698_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 228482_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 244698_at | - | - |
Human Genome U133 Plus 2.0 Array | 228482_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 244698_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 228482_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 244698_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 244698_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 244698_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228482_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 244698_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 244698_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228482_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228482_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228482_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 244698_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228482_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228482_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 244698_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 244698_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228482_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 244698_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228482_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 244698_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 244698_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228482_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228482_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 228482_at10 | - | - |
Human Genome U95 Set | 42021_PM_at | - | - |
Human Genome U95 Set | 42021_at | - | - |
Human Genome U95 Set | 77393_PM_at | - | - |
Human Genome U95 Set | 77393_at | - | - |
Human Genome U133 Set | 244698_PM_at | - | - |
Human Genome U133 Set | 228482_at | - | - |
Human Genome U133 Set | 228482_PM_at | - | - |
Human Genome U133 Set | 244698_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007967 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009221 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007967 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009221 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 185031 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 187667 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1691418 | AK172733.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1691418 | BM971397.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1691418 | NM_001204477.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1691418 | AK093993.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1691418 | AK172733.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1691418 | BM971397.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1691418 | NM_001204477.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1691418 | AK093993.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1691418 | AK172733.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1691418 | BM971397.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1691418 | NM_001204477.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1691418 | AK093993.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1691418 | AK172733.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1691418 | BM971397.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1691418 | NM_001204477.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1691418 | AK093993.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1691418 | AK172733.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1691418 | BM971397.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1691418 | NM_001204477.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1691418 | AK093993.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1691418 | AK172733.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1691418 | BM971397.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1691418 | NM_001204477.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1691418 | AK093993.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1691418 | AK172733.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1691418 | BM971397.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1691418 | NM_001204477.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1691418 | AK093993.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1691418 | AK172733.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1691418 | BM971397.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1691418 | NM_001204477.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1691418 | AK093993.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1691418 | AK172733.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1691418 | BM971397.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1691418 | NM_001204477.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1691418 | AK093993.1 | human_transcriptome |
