Assay Details
Gene Symbol: LOC729224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135243.1 | 3 | 132 | |||
NR_135244.1 | 3 | 132 | ||||
GenBank mRNA | BC040292.1 | - | 2 | 132 | ||
HY208135.1 | - | 2 | 338 | 132 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | KIAA2012 | KIAA2012 | Anti-sense | Chr. 2 - 202073245 - 202205195 on NCBI Build GRCh38 | ||
Downstream | small ubiquitin-like modifier 1 | SUMO1 |
DAP1, GMP1, OFC10, PIC1, SENP2, SMT3, SMT3C, SMT3H3, UBL1 |
91.6kb | Sense | Chr. 2 - 202206180 - 202238599 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942710 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942723 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817885 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942698 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942726 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942742 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817945 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817886 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817935 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942694 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817943 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817900 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817880 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817951 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942702 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942712 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817911 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942737 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817895 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817897 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942722 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817937 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817932 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942696 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942746 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817924 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942689 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817942 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817882 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942744 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942743 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942677 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942704 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817894 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817890 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817884 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817910 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942699 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817938 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817925 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817918 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942701 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942705 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817887 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942719 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817904 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817912 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942714 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817920 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817898 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942703 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942732 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817954 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817922 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21942724 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01817881 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3292148 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 14096 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3292148 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P16001780 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P22139677 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16001780 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1556536_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0500011353.hg.1 | ENST00000401306.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010760.hg.1 | ENST00000408477.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011681.hg.1 | ENST00000410702.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012112.hg.1 | ENST00000408510.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015443.hg.1 | ENST00000409819.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015443.hg.1 | ENST00000409819.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015443.hg.1 | ENST00000409819.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015443.hg.1 | ENST00000409819.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015443.hg.1 | ENST00000584624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015443.hg.1 | ENST00000584624.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013662.hg.1 | ENST00000408126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007681.hg.1 | ENST00000408114.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010354.hg.1 | ENST00000618854.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008489.hg.1 | ENST00000408372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010974.hg.1 | ENST00000408258.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011388.hg.1 | ENST00000390804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009271.hg.1 | ENST00000621256.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010401.hg.1 | ENST00000516992.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009809.hg.1 | ENST00000584402.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009809.hg.1 | NR_039699.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011681.hg.1 | ENST00000410702.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007681.hg.1 | ENST00000408114.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010760.hg.1 | ENST00000408477.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011388.hg.1 | ENST00000390804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009271.hg.1 | ENST00000621256.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500011353.hg.1 | ENST00000401306.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012112.hg.1 | ENST00000408510.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015443.hg.1 | ENST00000409819.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015443.hg.1 | ENST00000409819.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015443.hg.1 | ENST00000409819.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015443.hg.1 | ENST00000409819.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015443.hg.1 | ENST00000584624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015443.hg.1 | ENST00000584624.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008489.hg.1 | ENST00000408372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009809.hg.1 | ENST00000584402.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009809.hg.1 | NR_039699.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013662.hg.1 | ENST00000408126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010354.hg.1 | ENST00000618854.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010401.hg.1 | ENST00000516992.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010974.hg.1 | ENST00000408258.2 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1556536_at | - | - |
Human Genome U133 Plus 2.0 Array | 1556536_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556536_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556536_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556536_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556536_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556536_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556536_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556536_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556536_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556536_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556536_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556536_at8 | - | - |
Human Genome U95 Set | 68273_at | - | - |
Human Genome U95 Set | 68273_PM_at | - | - |
