Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148278.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148278.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148278.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148278.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148278.hg.1 |
TC0Y00007179.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0Y00148281.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148281.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148281.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148281.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148281.hg.1 |
TC0Y00007179.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0Y00007179.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148279.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148279.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148279.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148279.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148279.hg.1 |
TC0Y00007179.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0Y00148280.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148280.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148280.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148280.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148280.hg.1 |
TC0Y00007179.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0Y00148282.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148282.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148282.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148282.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148282.hg.1 |
TC0Y00007179.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0Y00148283.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148283.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148283.hg.1 |
ENST00000382764.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0Y00148283.hg.1 |
NM_001282471.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0Y00148283.hg.1 |
TC0Y00007179.hg.1 |
Clariom D TC |