Assay Details
Gene Symbol: ERICH6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121674.1 | 4 | 557 | 147 | ||
NR_121675.1 | 3 | 328 | 147 | |||
GenBank mRNA | DB324976.1 | - | 1 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012805.hg.1 | ENST00000474888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012805.hg.1 | ENST00000474888.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165820.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165820.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165820.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165820.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165820.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165820.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165820.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165820.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165820.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165820.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165820.hg.1 | TC0300009171.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300012805.hg.1 | ENST00000474888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012805.hg.1 | ENST00000474888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009171.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165817.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165817.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165817.hg.1 | TC0300009171.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300165819.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165819.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165819.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165819.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165819.hg.1 | TC0300009171.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300165825.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165825.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165825.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165825.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165825.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165825.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165825.hg.1 | TC0300009171.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300165816.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165816.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165816.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165816.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165816.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165816.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165816.hg.1 | TC0300009171.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165821.hg.1 | TC0300009171.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | ENST00000463755.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | ENST00000471093.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | ENST00000475393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | NR_121674.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | NR_121675.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300165823.hg.1 | TC0300009171.hg.1 | Clariom D TC |
