Assay Details
Gene Symbol: ERICH6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121674.1 | 4 | 580 | 91 | ||
NR_121675.1 | 3 | 351 | 91 | |||
GenBank mRNA | DB324976.1 | - | 1 | 91 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
