Assay Details
| Gene Symbol: EIF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300821.1 | NP_001287750.1 | 15 | 2358 | 154 | |
| NM_001417.5 | NP_001408.2 | 15 | 2343 | 154 | ||
| GenBank mRNA | AK299510.1 | - | 11 | 1417 | 154 | |
| AK307496.1 | - | 5 | 772 | 154 | ||
| BC001097.1 | - | 8 | 1332 | 154 | ||
| BC073139.1 | - | 15 | 2133 | 6527540 | 154 | |
| CR627377.1 | - | 9 | 1202 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

EIF4B
Genomic Map