Assay Details
| Gene Symbol: FLT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002019.4 | NP_002010.2 | 30 | 4687 | 105 | |
| XM_017020485.1 | XP_016875974.1 | 29 | 4573 | 105 | ||
| GenBank mRNA | CA396081.1 | - | 1 | 159 | 105 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FLT1
Genomic Map