Assay Details
Gene Symbol: LHPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167880.1 | NP_001161352.1 | 6 | 1202 | 95 | |
NM_001318331.1 | NP_001305260.1 | 4 | 1045 | 95 | ||
NM_022126.3 | NP_071409.3 | 7 | 1294 | 95 | ||
XM_006717943.1 | XP_006718006.1 | 8 | 1315 | 95 | ||
GenBank mRNA | AK055532.1 | - | 7 | 1223 | 95 | |
BC110344.1 | - | 6 | 1156 | 5762731 | 95 | |
BC113629.1 | - | 8 | 1232 | 8322681 | 95 | |
BC113631.1 | - | 8 | 1215 | 8322683 | 95 | |
EF151005.1 | - | 2 | 802 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000905.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000481452.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000481452.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000481452.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000481452.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000481452.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000482963.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000482963.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000482963.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000486535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000486535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000486535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000486535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000487190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000487190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000487190.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000492187.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000492187.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000492187.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000493240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000493240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000493240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | ENST00000493240.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10011089.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011089.hg.1 | ENST00000368842.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10011089.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10011089.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10011089.hg.1 | TC10000905.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000186355.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186355.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186355.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186355.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186355.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186355.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000186355.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186355.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000186355.hg.1 | TC1000012505.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368839.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000368842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000392757.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000481452.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000481452.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000481452.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000481452.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000481452.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000482963.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000482963.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000482963.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000486535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000486535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000486535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000486535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000487190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000487190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000487190.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000492187.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000492187.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000492187.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000493240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000493240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000493240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | ENST00000493240.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_001167880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012505.hg.1 | NM_022126.3 | Refseq |
