| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008103.hg.1 |
ENST00000522365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008103.hg.1 |
ENST00000522365.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008103.hg.1 |
NR_122034.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800008103.hg.1 |
NR_122034.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155585.hg.1 |
ENST00000522365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155585.hg.1 |
NR_122034.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155585.hg.1 |
ENST00000522365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155585.hg.1 |
NR_122034.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155585.hg.1 |
TC0800008103.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800155583.hg.1 |
ENST00000522365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155583.hg.1 |
NR_122034.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155583.hg.1 |
ENST00000522365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800155583.hg.1 |
NR_122034.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800155583.hg.1 |
TC0800008103.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800008103.hg.1 |
ENST00000522365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008103.hg.1 |
ENST00000522365.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800008103.hg.1 |
NR_122034.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800008103.hg.1 |
NR_122034.1 |
Refseq |