Assay Details
Gene Symbol: LOC100505534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL137606.1 | - | 1 | 3249 | 96 | |
BC042903.1 | - | 2 | 2149 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | G protein-coupled receptor 176 | GPR176 | HB-954 | -1.9kb | Anti-sense | Chr. 15 - 39795049 - 39920933 on NCBI Build GRCh38 |
Downstream | eukaryotic translation initiation factor 2 alpha kinase 4 | EIF2AK4 |
GCN2, PVOD2 |
9.2kb | Sense | Chr. 15 - 39934101 - 40035596 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700010912.hg.1 | ENST00000458981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400011630.hg.1 | ENST00000365666.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300007154.hg.1 | ENST00000362918.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009079.hg.1 | ENST00000362539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009438.hg.1 | ENST00000362353.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011480.hg.1 | ENST00000364992.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400008878.hg.1 | ENST00000363179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008523.hg.1 | ENST00000384395.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100016584.hg.1 | ENST00000384178.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009722.hg.1 | ENST00000362574.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006929.hg.1 | ENST00000499797.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006929.hg.1 | ENST00000499797.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006929.hg.1 | ENST00000499797.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006929.hg.1 | ENST00000499797.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007030.hg.1 | ENST00000459189.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010953.hg.1 | ENST00000364572.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016158.hg.1 | ENST00000384322.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008983.hg.1 | ENST00000384768.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011266.hg.1 | ENST00000365131.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008985.hg.1 | ENST00000363444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008162.hg.1 | ENST00000384078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009516.hg.1 | ENST00000364439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011417.hg.1 | ENST00000365201.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006972.hg.1 | ENST00000362352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011677.hg.1 | ENST00000384063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012316.hg.1 | ENST00000365439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200011508.hg.1 | ENST00000363025.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008170.hg.1 | ENST00000384657.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006839.hg.1 | ENST00000384131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007064.hg.1 | ENST00000363832.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009075.hg.1 | ENST00000363435.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007561.hg.1 | ENST00000384494.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007131.hg.1 | ENST00000363549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015884.hg.1 | ENST00000384312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011666.hg.1 | ENST00000362492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500012038.hg.1 | ENST00000362807.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010816.hg.1 | ENST00000384563.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008223.hg.1 | ENST00000363294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008419.hg.1 | ENST00000384670.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700011492.hg.1 | ENST00000365165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014983.hg.1 | ENST00000364218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700008055.hg.1 | ENST00000364268.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400008878.hg.1 | ENST00000363179.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010816.hg.1 | ENST00000384563.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500012038.hg.1 | ENST00000362807.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011677.hg.1 | ENST00000384063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015884.hg.1 | ENST00000384312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009079.hg.1 | ENST00000362539.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008985.hg.1 | ENST00000363444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011480.hg.1 | ENST00000364992.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007030.hg.1 | ENST00000459189.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007561.hg.1 | ENST00000384494.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008523.hg.1 | ENST00000384395.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016158.hg.1 | ENST00000384322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009516.hg.1 | ENST00000364439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700011492.hg.1 | ENST00000365165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011666.hg.1 | ENST00000362492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008162.hg.1 | ENST00000384078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009722.hg.1 | ENST00000362574.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014983.hg.1 | ENST00000364218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010953.hg.1 | ENST00000364572.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800011266.hg.1 | ENST00000365131.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200011508.hg.1 | ENST00000363025.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011417.hg.1 | ENST00000365201.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008223.hg.1 | ENST00000363294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012316.hg.1 | ENST00000365439.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008419.hg.1 | ENST00000384670.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006972.hg.1 | ENST00000362352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300007154.hg.1 | ENST00000362918.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006929.hg.1 | ENST00000499797.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006929.hg.1 | ENST00000499797.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006929.hg.1 | ENST00000499797.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006929.hg.1 | ENST00000499797.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006929.hg.1 | ENST00000558675.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009075.hg.1 | ENST00000363435.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009438.hg.1 | ENST00000362353.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100016584.hg.1 | ENST00000384178.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400011630.hg.1 | ENST00000365666.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008983.hg.1 | ENST00000384768.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007064.hg.1 | ENST00000363832.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700008055.hg.1 | ENST00000364268.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006839.hg.1 | ENST00000384131.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008170.hg.1 | ENST00000384657.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007131.hg.1 | ENST00000363549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010912.hg.1 | ENST00000458981.1 | Ensembl |
