Assay Details
Gene Symbol: LINC01568 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC042734.1 | - | 2 | 2471 | 134 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000554640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000555721.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000561875.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | ENST00000562661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600008384.hg.1 | NR_038234.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 689853 | - | - |
