Assay Details
| Gene Symbol: LINC01372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108104.1 | 3 | 422 | 86 | ||
| GenBank mRNA | BC017910.1 | - | 3 | 423 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LINC01372
Genomic Map