Assay Details
Gene Symbol: LINC01597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK127732.1 | - | 2 | 1744 | 102 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11129613 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31776169 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11129611 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17185639 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11129612 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31776130 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17185640 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 28004 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3340094 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P41302835 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03505165 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 69407 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P840728 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8065548 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8065549 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8065547 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000532103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000532103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000532103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000532103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000532103.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000532731.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000528459.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000532103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000532103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000532103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000532103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000532103.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000532731.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012999.hg.1 | ENST00000533230.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1797431 | AK127732.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1797431 | AK127732.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1797431 | AK127732.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1797431 | AK127732.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1797431 | AK127732.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1797431 | AK127732.1 | human_transcriptome |
