Assay Details
| Gene Symbol: FANCD2OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173472.1 | NP_775743.1 | 3 | 849 | 143 | |
| GenBank mRNA | BC028122.1 | - | 3 | 849 | 5168387 | 143 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001157.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03019982.hg.1 | TC03001157.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019982.hg.1 | ENST00000436517.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019982.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019982.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000450616.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000450616.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000450616.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000453223.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000453223.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019991.hg.1 | TC03001157.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019991.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019991.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019991.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019991.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000450616.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000450616.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000450616.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000453223.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000453223.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010228.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300174626.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174626.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174626.hg.1 | ENST00000450616.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174626.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174626.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300174626.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174626.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174626.hg.1 | ENST00000450616.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174626.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174626.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300174626.hg.1 | TC0300010228.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300174627.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174627.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174627.hg.1 | ENST00000450616.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174627.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174627.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300174627.hg.1 | ENST00000431315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174627.hg.1 | ENST00000436517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174627.hg.1 | ENST00000450616.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174627.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174627.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300174627.hg.1 | TC0300010228.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300174645.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174645.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174645.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300174645.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300174645.hg.1 | ENST00000450660.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174645.hg.1 | ENST00000524279.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174645.hg.1 | NM_001164839.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300174645.hg.1 | NM_173472.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300174645.hg.1 | TC0300010228.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 162476 | - | - |

FANCD2OS
Genomic Map