Assay Details
| Gene Symbol: RNASE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002933.4 | NP_002924.1 | 2 | 692 | 159 | |
| NM_198232.2 | NP_937875.1 | 2 | 762 | 159 | ||
| NM_198234.2 | NP_937877.1 | 3 | 763 | 159 | ||
| NM_198235.2 | NP_937878.1 | 3 | 775 | 159 | ||
| GenBank mRNA | AK098375.1 | - | 2 | 742 | 159 | |
| AK129871.1 | - | 2 | 683 | 159 | ||
| BC005324.1 | - | 2 | 692 | 3950230 | 159 | |
| BC022882.1 | - | 2 | 666 | 4294153 | 159 | |
| BM919747.1 | - | 2 | 647 | 159 | ||
| BP385178.1 | - | 1 | 454 | 159 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000902.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009768.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009768.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009768.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009768.hg.1 | ENST00000555698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009768.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009768.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009768.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009768.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14009768.hg.1 | TC14000902.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400161908.hg.1 | TC1400008606.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000340900.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397967.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000397970.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000412779.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | ENST00000555698.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_002933.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198232.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198234.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008606.hg.1 | NM_198235.2 | Refseq |

RNASE1
Genomic Map