Assay Details
Gene Symbol: OR10Z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004478.1 | NP_001004478.1 | 1 | 821 | 123 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001344.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
G-protein coupled receptor activity
olfactory receptor activity
G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
G-protein coupled receptor activity
olfactory receptor activity
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537274 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596067 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537270 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537259 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537281 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596057 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596080 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537285 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537264 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596055 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596076 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596058 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537267 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596078 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537268 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537272 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596071 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596064 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15135156 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596054 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596079 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596056 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537279 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537271 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596070 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537265 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20596081 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00537282 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3381107 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0393 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0391 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0392 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12081 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P00173032 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3381107 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3381107 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537285 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596056 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596054 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596055 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596067 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596058 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596071 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596070 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596079 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537279 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596076 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596078 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596064 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537274 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537272 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537265 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537264 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537259 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537264 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537272 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537279 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537285 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596064 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537265 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596078 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596055 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596076 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596079 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596067 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537274 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596070 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596058 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00537259 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596071 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596054 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20596056 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537272 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537277 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537287 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537264 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537276 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537265 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537281 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537261 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537259 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537284 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537268 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537269 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537285 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537274 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00537263 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537277 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537284 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537265 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537261 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15135155 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537281 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537272 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537267 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537263 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537285 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537259 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15135157 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537289 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537287 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537276 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537274 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00537264 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001004478_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7906373 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7906372 | - | - |
Affymetrix Human Genome U219 Array | 11742581_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11742581_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0100010238.hg.1 | ENST00000361284.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010238.hg.1 | NM_001004478.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01021072.hg.1 | TC01001344.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01021072.hg.1 | ENST00000361284.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01021072.hg.1 | NM_001004478.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010238.hg.1 | ENST00000361284.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010238.hg.1 | NM_001004478.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100175096.hg.1 | ENST00000361284.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100175096.hg.1 | NM_001004478.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100175096.hg.1 | ENST00000361284.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100175096.hg.1 | NM_001004478.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100175096.hg.1 | TC0100010238.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033039 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033039 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1797393 | NM_001004478.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1797393 | NM_001004478.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1797393 | NM_001004478.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1797393 | NM_001004478.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1797393 | NM_001004478.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_27126-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1797393 | NM_001004478.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1797393 | NM_001004478.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 2450408 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_27126 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1797393 | NM_001004478.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1797393 | NM_001004478.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1797393 | NM_001004478.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1797393 | NM_001004478.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1797393 | NM_001004478.1 | human_transcriptome |
