Assay Details
Gene Symbol: OR2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004687.1 | NP_001004687.1 | 1 | 343 | 140 | |
GenBank mRNA | BC137039.1 | - | 1 | 363 | 9021035 | 140 |
BC137040.1 | - | 1 | 363 | 9021036 | 140 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002028.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005583 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931337 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005592 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931332 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931334 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005587 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005580 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005582 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21005589 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931333 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931330 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15205214 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3369975 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0420 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0419 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 30094 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3370001 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3369975 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3369975 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931333 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005589 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005591 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931337 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931334 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005582 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005593 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931330 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205214 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005583 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005587 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931330 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931333 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931337 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005587 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005582 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005583 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005593 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205214 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005589 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21005591 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931334 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931336 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931333 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931339 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931338 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931334 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931331 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931341 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931333 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931338 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931341 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931336 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931327 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931329 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931332 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001004687_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7911259 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7911258 | - | - |
Affymetrix Human Genome U219 Array | 11759267_at | - | - |
Affymetrix Human Genome U219 Array | 11759267_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0100012345.hg.1 | ENST00000359959.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012345.hg.1 | NM_001004687.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01031117.hg.1 | TC01002028.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01031117.hg.1 | ENST00000359959.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01031117.hg.1 | NM_001004687.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012334.hg.1 | ENST00000357191.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012334.hg.1 | ENST00000623922.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012334.hg.1 | NM_001001963.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190608.hg.1 | ENST00000359959.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190608.hg.1 | NM_001004687.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190608.hg.1 | ENST00000359959.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190608.hg.1 | NM_001004687.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190608.hg.1 | TC0100012345.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100012334.hg.1 | ENST00000357191.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012334.hg.1 | ENST00000623922.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012334.hg.1 | NM_001001963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012345.hg.1 | ENST00000359959.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012345.hg.1 | NM_001004687.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042316 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042316 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1754846 | NM_001004687.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1754846 | BC137039.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1754846 | BC137040.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1754846 | NM_001004687.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1754846 | BC137039.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1754846 | BC137040.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1754846 | NM_001004687.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1754846 | BC137039.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1754846 | BC137040.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1754846 | NM_001004687.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1754846 | BC137039.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1754846 | BC137040.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1754846 | NM_001004687.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1754846 | BC137039.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1754846 | BC137040.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_23254-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1790301 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1754846 | NM_001004687.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1754846 | BC137039.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1754846 | BC137040.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1790301 | - | - |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1754846 | NM_001004687.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1754846 | BC137039.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1754846 | BC137040.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_23254 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_23254 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1754846 | NM_001004687.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1754846 | BC137039.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1754846 | BC137040.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1754846 | NM_001004687.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1754846 | BC137039.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1754846 | BC137040.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1754846 | NM_001004687.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1754846 | BC137039.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1754846 | BC137040.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1754846 | NM_001004687.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1754846 | BC137039.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1754846 | BC137040.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1754846 | NM_001004687.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1754846 | BC137039.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1754846 | BC137040.1 | human_transcriptome |
