Assay Details
Gene Symbol: OR5M10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004741.1 | NP_001004741.1 | 1 | 440 | 127 | |
GenBank mRNA | BC140733.1 | - | 1 | 506 | 9021601 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219552 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219527 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219556 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219557 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219558 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219542 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219561 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809968 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219525 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219529 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219544 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219522 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219537 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809941 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219530 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219531 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219549 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219541 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219562 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219535 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809951 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219526 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219536 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219554 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219523 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809960 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219547 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219559 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809978 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219550 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28219521 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809955 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07809983 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3253299 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0474 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0473 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 16590 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3253299 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3343496 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3253299 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219521 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809955 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219527 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219525 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219529 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809941 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809942 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809951 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809960 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219541 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219542 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219537 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219535 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219556 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219562 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809983 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219530 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219531 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219544 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219559 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219526 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219550 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809956 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16453544 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219523 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219549 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219552 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219531 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809956 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809960 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219536 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219541 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219562 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219523 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219526 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809983 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219525 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809955 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809942 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219554 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809941 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219542 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219552 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219550 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809951 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219521 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219527 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07809968 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16453544 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219530 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219549 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219544 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219535 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219522 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219557 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219559 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219529 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219556 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28219537 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809962 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809971 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809964 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809973 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809978 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809977 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809941 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809981 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809957 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809986 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809969 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809959 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809952 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809950 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809944 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809954 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809983 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809966 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809948 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809946 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07809975 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001004741_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7948149 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7948148 | - | - |
Affymetrix Human Genome U219 Array | 11742354_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11742354_at | - | - |
Affymetrix Human Clariom D Assay | PSR1100182129.hg.1 | TC1100010864.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045173 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045173 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1710884 | NM_001004741.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1710884 | BC140733.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2136737 | NM_001004741.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2136737 | BC140733.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1710884 | NM_001004741.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1710884 | BC140733.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1710884 | NM_001004741.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1710884 | BC140733.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2136737 | NM_001004741.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2136737 | BC140733.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2136737 | NM_001004741.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2136737 | BC140733.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2136737 | NM_001004741.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2136737 | BC140733.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_30216-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1710884 | NM_001004741.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1710884 | BC140733.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1710884 | NM_001004741.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1710884 | BC140733.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 4250066 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_30216 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2136737 | NM_001004741.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2136737 | BC140733.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2136737 | NM_001004741.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2136737 | BC140733.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1710884 | NM_001004741.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1710884 | BC140733.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1710884 | NM_001004741.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1710884 | BC140733.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2136737 | NM_001004741.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2136737 | BC140733.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1710884 | NM_001004741.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1710884 | BC140733.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2136737 | NM_001004741.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2136737 | BC140733.1 | human_transcriptome |
